Key Documents
Z100579
Sigma-Aldrich
Single-bulb ball-tube
capacity 25 mL, joint: ST/NS 14/20, plain
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Pricing and availability is not currently available.
Joint
joint: ST/NS14/20
capacity
25 mL
General description
Arranged horizontally outside of Kugelrohr oven, the ball-tubes cool and collect distillate with minimal loss of material.
Choose from one of the most recent versions:
Certificates of Analysis (COA)