Key Documents
WHA10342581
Whatman® Seed testing paper
L × W (420 mm) (594 mm), pack of 50 ea
About This Item
UNSPSC Code:
41115727
NACRES:
NB.24
Pricing and availability is not currently available.
packaging
pack of 50 ea
manufacturer/tradename
Cytiva
Whatman 10342581
L × W
(420 mm) (594 mm)
Related Categories
General description
Owing to their extremely high purity Whatman seed testing papers provide reliable and reproducible results. The papers are made from pure cellulose without any additives and do not contain any substances which could influence the growth of the seeds. The constant water absorption of the papers ensures the continuous provision of the required amount of water.
The better contrast of the color seed testing papers makes evaluation easier, particularly for seeds with fine white rootlets or under artificial light. This makes work easier, improves the results and saves time. The dyes which we use have been thoroughly investigated and have no influence on the growth of the seeds.
Features and Benefits
  • The constant water absorption of the papers ensures the continuous provision of the required amount of water.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)