Skip to Content
Merck
All Photos(1)

Key Documents

W370401

Sigma-Aldrich

4-Methyl-2,6-dimethoxyphenol

≥97%

Synonym(s):

2,6-dimethoxy-p-cresol, 3,5-dimethoxy-4-hydroxytoluene, 4-methylsyringol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H2(OCH3)2OH
CAS Number:
Molecular Weight:
168.19
FEMA Number:
3704
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.053
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Assay

≥97%

bp

145-146 °C/14 mmHg (lit.)

mp

37-42 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

medicinal; smoky; spicy; woody; phenolic

SMILES string

COc1cc(C)cc(OC)c1O

InChI

1S/C9H12O3/c1-6-4-7(11-2)9(10)8(5-6)12-3/h4-5,10H,1-3H3

InChI key

ZFBNNSOJNZBLLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service