Skip to Content
Merck
All Photos(1)

Key Documents

W358401

Sigma-Aldrich

1-Penten-3-ol

≥98%, FG

Synonym(s):

Ethyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(OH)CH=CH2
CAS Number:
Molecular Weight:
86.13
FEMA Number:
3584
Beilstein:
1719833
EC Number:
Council of Europe no.:
4025
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.099
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.424 (lit.)

bp

114-115 °C (lit.)

density

0.838 g/mL at 20 °C (lit.)
0.839 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; horseradish; fruity; vegetable; pungent; tropical

SMILES string

CCC(O)C=C

InChI

1S/C5H10O/c1-3-5(6)4-2/h3,5-6H,1,4H2,2H3

InChI key

VHVMXWZXFBOANQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • A Novel Strategy for Mixed Jam Evaluation: Apparent Indicator, Sensory, Metabolomic, and GC-IMS Analysis.: This study demonstrates the use of 1-Penten-3-ol in enhancing the sensory profiles of mixed fruit jams. The research utilizes gas chromatography-ion mobility spectrometry (GC-IMS) for detailed volatile compound analysis, highlighting its utility in food science for flavor enhancement and quality control (Maimaitiyiming et al., 2024).

  • Extending the Shelf Life of Atlantic Salmon (Salmo salar) with Sub-Chilled Storage and Modified Atmosphere Packaging in Recyclable Mono-Material Trays.: This paper explores the impact of 1-Penten-3-ol in modified atmosphere packaging, focusing on its efficacy in prolonging the freshness of seafood. The study provides insights into sustainable packaging solutions and food preservation, crucial for reducing food waste and enhancing food safety (Chan et al., 2023).

  • Flavor and Sensory Profile of Chinese Traditional Fish Noodles Produced by Different Silver Carp (hypophthalmichthys molitrix) Mince Ingredients.: Research into the use of 1-Penten-3-ol in enhancing the flavor profile of traditional Chinese fish noodles, emphasizing its role in improving consumer acceptance and culinary innovation. The study combines sensory analysis and flavor profiling, offering valuable data for food chemists and technologists (Zhou et al., 2023).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A M Killary et al.
Experimental cell research, 154(2), 442-453 (1984-10-01)
A selective system toxic towards mammalian cells expressing the liver-specific isozyme of alcohol dehydrogenase (L-ADH) has been developed. A number of alpha-unsaturated primary and secondary alcohols were assayed for their ability to serve as substrates for rat liver ADH and
A J Fisher et al.
Phytochemistry, 62(2), 159-163 (2002-12-17)
Large releases of 1-penten-3-ol (pentenol) and 1-penten-3-one (pentenone) were recently observed from a variety of leaves subjected to freeze-thaw damage in the presence of oxygen. In order to understand the biochemical origins of these volatiles, soybean leaf extracts were used
J J Garrido et al.
Biochemical genetics, 28(9-10), 513-522 (1990-10-01)
The detoxification of 1-pentene-3-ol (pentenol) and 1-pentene-3-one (pentenone) by Drosophila melanogaster adult flies has been studied in two homozygous lines for the AdhF and AdhS alleles (LRC lines), in their respective lines selected for tolerance to ethanol (LRSe lines) and
J J Garrido et al.
Heredity, 61 ( Pt 1), 85-91 (1988-08-01)
The participation of the ADH enzymes in the detoxification by D. melanogaster of 1-pentene-3-ol (also called pentenol) and its oxidized product, 1-pentene-3-one (usually known as ethyl-vinyl-ketone or pentenone) have been studied using the LR lines. For this purpose flies of
Ana Rivas-Cañedo et al.
Meat science, 93(2), 178-186 (2012-10-03)
The effect of dietary supplementation with either vitamin E (300 ppm) or a red wine extract rich in polyphenols (900 ppm) in an omega-3 enriched concentrate on the volatile fraction of lamb meat was assessed. The effect of refrigerated storage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service