Skip to Content
Merck
All Photos(1)

Key Documents

W340707

Sigma-Aldrich

trans-2-Methyl-2-butenal

sum of isomers, ≥99%, FG

Synonym(s):

Tiglic aldehyde, (E)-2-Methylbut-2-enal, trans-2,3-Dimethylacrolein, trans-2-Methyl-2-butenal, Tiglinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)CHO
CAS Number:
Molecular Weight:
84.12
FEMA Number:
3407
Beilstein:
1698207
Council of Europe no.:
2281
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.095
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥99%

impurities

<2.0% tiglic acid

refractive index

n20/D 1.448 (lit.)

bp

116-119 °C/752 mmHg (lit.)
116-122 °C

density

0.869 g/mL at 20 °C
0.871 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; fruity

SMILES string

C\C=C(/C)C=O

InChI

1S/C5H8O/c1-3-5(2)4-6/h3-4H,1-2H3/b5-3+

InChI key

ACWQBUSCFPJUPN-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-2-Methyl-2-butenal is one of the major volatile constituents of coriander and olive oil.

Application


  • Gas-Phase Reaction of trans-2-Methyl-2-butenal with Cl: Kinetics, Gaseous Products, and SOA Formation.: This study provides a comprehensive analysis of the atmospheric reactions involving trans-2-Methyl-2-butenal, highlighting its impact on air quality and secondary organic aerosol (SOA) formation. Such insights are crucial for environmental chemists and policymakers aiming to understand and mitigate air pollution (Antiñolo et al., 2020).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterisation of 39 varietal virgin olive oils by their volatile compositions.
Luna G, et al.
Food Chemistry, 98(2), 243-252 (2006)
Free and glycosidically bound volatiles in coriander herb (Coriandrum sativum).
Padmakumari KP.
Journal of Essential Oil-Bearing Plants, 11(6), 615-622 (2008)
Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service