Skip to Content
Merck
All Photos(1)

Key Documents

W316255

Sigma-Aldrich

Furfuryl thioacetate

natural, ≥98%, FG

Synonym(s):

S-Furfuryl thioacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2S
CAS Number:
Molecular Weight:
156.20
FEMA Number:
3162
MDL number:
UNSPSC Code:
12164502
Flavis number:
13.033
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
botanical
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

botanical

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

burnt; coffee; meaty; roasted

storage temp.

2-8°C

SMILES string

S(Cc1[o]ccc1)C(=O)C

InChI

1S/C7H8O2S/c1-6(8)10-5-7-3-2-4-9-7/h2-4H,5H2,1H3

InChI key

LQOUTUIIYXYBQW-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service