Skip to Content
Merck
All Photos(1)

Key Documents

W285706

Sigma-Aldrich

Phenethyl acetate

≥98%, FCC, FG

Synonym(s):

2-Phenethyl acetate, 2-Phenylethyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2C6H5
CAS Number:
Molecular Weight:
164.20
FEMA Number:
2857
Beilstein:
638179
EC Number:
Council of Europe no.:
221
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.031
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor density

5.67 (vs air)

Assay

≥98%

refractive index

n20/D 1.498 (lit.)

bp

238-239 °C (lit.)

density

1.032 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

honey; floral; rose; sweet

SMILES string

CC(=O)OCCc1ccccc1

InChI

1S/C10H12O2/c1-9(11)12-8-7-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3

InChI key

MDHYEMXUFSJLGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phenethyl acetate is a volatile flavor compound reported to be found in Malaysian cocoa beans, cheddar cheese, wine, brandy, and other grape-derived alcoholic beverages.

Biochem/physiol Actions

Taste at 50 ppm

Other Notes

Natural occurrence: Melon, pineapple, grape, brandy, Arctic bramble, olives and lettuce.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of aroma compounds responsible for the rosy/floral flavor in Cheddar cheese.
Carunchia Whetstine ME, et al.
Journal of Agricultural and Food Chemistry, 53(8), 3126-3132 (2005)
Effect of increased yeast alcohol acetyltransferase activity on flavor profiles of wine and distillates.
Lilly M, et al.
Applied and Environmental Microbiology, 66(2), 744-753 (2000)
Philipp Adler et al.
Enzyme and microbial technology, 48(3), 285-292 (2011-11-25)
An unstructured model for an integrated fermentation/membrane extraction process for the production of the aroma compounds 2-phenylethanol and 2-phenylethylacetate by Kluyveromyces marxianus CBS 600 was developed. The extent to which this model, based only on data from the conventional fermentation
Patrick C Aculey et al.
Journal of food science, 75(6), S300-S307 (2010-08-21)
Export of cocoa beans is of great economic importance in Ghana and several other tropical countries. Raw cocoa has an astringent, unpleasant taste, and flavor, and has to be fermented, dried, and roasted to obtain the characteristic cocoa flavor and
Fernando Viana et al.
International journal of food microbiology, 135(1), 68-74 (2009-08-18)
The impact of mixed cultures of Hanseniaspora osmophila and Saccharomyces cerevisiae with different initial yeast ratios on wine composition has been examined. The mixed culture significantly affected sugar consumption, the main enological parameters and ester concentrations, with the exception of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service