Skip to Content
Merck
All Photos(1)

Key Documents

W263907

Sigma-Aldrich

Linalyl butyrate

≥95%, FG

Synonym(s):

3,7-dimethyl-1,6-octadien-3-yl butyrate, 3,7-dimethylocta-1,6-dien-3-yl butanoate, Butyric acid linalyl ester, Linalool butyrate, Linalyl 3-methylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2C(CH=CH2)(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
224.34
FEMA Number:
2639
EC Number:
Council of Europe no.:
276
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.050
NACRES:
NA.21
Organoleptic:
berry; fruity; floral; sweet
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.45 (lit.)

bp

80-82 °C/0.2 mmHg (lit.)

density

0.892 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

berry; fruity; floral; sweet

SMILES string

CCCC(=O)OC(C)(CC\C=C(\C)C)C=C

InChI

1S/C14H24O2/c1-6-9-13(15)16-14(5,7-2)11-8-10-12(3)4/h7,10H,2,6,8-9,11H2,1,3-5H3

InChI key

FHLGUOHLUFIAAA-UHFFFAOYSA-N

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C S Letizia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(7), 983-987 (2003-06-14)
A toxicologic and dermatologic review of linalyl butyrate when used as a fragrance ingredient is presented.
Polina D Blagojević et al.
Chemistry & biodiversity, 12(8), 1237-1255 (2015-08-13)
The plant volatile profile and the essential-oil chemical composition change during the storage of plant material. The objective of this study was to develop a mathematical model able to predict, explain, and quantify these changes. Mathematical equations, derived under the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service