Skip to Content
Merck
All Photos(1)

Key Documents

W221811

Sigma-Aldrich

Butyl isovalerate

natural, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COO(CH2)3CH3
CAS Number:
Molecular Weight:
158.24
FEMA Number:
2218
Beilstein:
1752803
EC Number:
Council of Europe no.:
444
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.449
NACRES:
NA.21
grade:
FG
Fragrance grade
Kosher
natural
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

FG
Fragrance grade
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

98%

refractive index

n20/D 1.409 (lit.)

bp

175 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; green; fruity; peach; pineapple; sweet

SMILES string

CCCCOC(=O)CC(C)C

InChI

1S/C9H18O2/c1-4-5-6-11-9(10)7-8(2)3/h8H,4-7H2,1-3H3

InChI key

AYWJSCLAAPJZEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service