Skip to Content
Merck
All Photos(1)

Key Documents

W221104

Sigma-Aldrich

Butyl propionate

≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CO2(CH2)3CH3
CAS Number:
Molecular Weight:
130.18
FEMA Number:
2211
EC Number:
Council of Europe no.:
405
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.124
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

4.5 (vs air)

Assay

≥98%

autoignition temp.

799 °F

refractive index

n20/D 1.401 (lit.)

bp

145 °C/756 mmHg (lit.)

density

0.875 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

banana; ethereal

SMILES string

CCCCOC(=O)CC

InChI

1S/C7H14O2/c1-3-5-6-9-7(8)4-2/h3-6H2,1-2H3

InChI key

BTMVHUNTONAYDX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F

Flash Point(C)

38 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M I Banton et al.
Journal of toxicology and environmental health. Part A, 61(2), 79-105 (2000-10-14)
Two inhalation studies were conducted to evaluate the possible subchronic and developmental toxic effects of n-butyl propionate. In the subchronic study, Sprague-Dawley rats (15/sex/group) were exposed to 0, 250, 750, or 1500 ppm vapor for 6 h/d, 5 d/wk for
Peng Liang et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 11(18), 4097-4102 (2010-08-19)
Rate coefficients for the gas-phase reactions of OH radicals with n-propyl butyrate (k(1)), n-butyl propionate (k(2)) and n-butyl butyrate (k(3)) are measured by both absolute and relative methods. The kinetics data obtained over the temperature range 273-372 K are used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service