Key Documents
SYND272440
Synthware distillation distribution adapter
joint: ST/NS 24/40
Synonym(s):
Synthware D271440, short path distillation receiver
About This Item
UNSPSC Code:
41104812
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware D272440
Related Categories
General description
With the outer and inner joint at 135° angle, used with short-path.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)