Skip to Content
Merck
All Photos(1)

Key Documents

QBD10260

Sigma-Aldrich

m-dPEG®8-NHS ester

Synonym(s):

2,5,8,11,14,17,20,23-Octaoxahexacosan-26-oic acid succinimidyl ester, 2,5-Dioxo-1-pyrrolidinyl 4,7,10,13,16,19,22,25-octaoxahexacosanoate, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H39NO12
CAS Number:
Molecular Weight:
509.54
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chemical modification reagent
reagent type: cross-linking reagent
reactivity: amine reactive

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCOCCOCCOCCOCCOCCOCCOCCOC)ON1C(CCC1=O)=O

InChI

1S/C22H39NO12/c1-27-6-7-29-10-11-31-14-15-33-18-19-34-17-16-32-13-12-30-9-8-28-5-4-22(26)35-23-20(24)2-3-21(23)25/h2-19H2,1H3

InChI key

IBZNTYBFTKFSMU-UHFFFAOYSA-N

Features and Benefits

The m-dPEG®8-NHS ester provides a variety of means to modify small molecules, peptides, proteins, and functionalized surfaces (metals, glass, and others). The single molecular weight methoxy-terminated dPEG® molecules are hydrophilic and non-immunogenic of exact length. Modification with m-dPEG® reagents can improve water solubility, increase hydrodynamic volume, eliminate or reduce non-specific binding, protect peptides and proteins from proteolysis, reduce or eliminate immunogenicity, and increase serum half-life in vivo. The m-dPEG®8-NHS ester reacts with amines.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service