Skip to Content
Merck
All Photos(4)

Key Documents

P6395

Sigma-Aldrich

Sodium phosphotungstate octadecahydrate

Synonym(s):

Tungstophosphoric acid sodium salt octadecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
∼ 2Na2O·P2O5·12WO3·18H2O
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

powder

reaction suitability

reagent type: catalyst
core: tungsten

SMILES string

[Na+].[Na+].[Na+].O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.[O-]P([O-])([O-])=O

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Azua-Bustos et al.
Scientific reports, 8(1), 16706-16706 (2018-11-14)
The hyperarid core of the Atacama Desert, the driest and oldest desert on Earth, has experienced a number of highly unusual rain events over the past three years, resulting in the formation of previously unrecorded hypersaline lagoons, which have lasted
Victoria DeJarnatt et al.
Journal of histotechnology, 44(1), 37-45 (2020-11-12)
Neutral-buffered formalin (NBF) has been used as the primary fixative in anatomic pathology laboratories for decades. Although it yields excellent morphologic and staining results, NBF poses significant health hazards requiring tissue to be grossed under a grossing/chemical fume hood. Glyoxal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service