Skip to Content
Merck
All Photos(1)

Key Documents

M4007

Sigma-Aldrich

2,2′-Thiodiethanethiol

technical grade, 90%

Synonym(s):

2-Mercaptoethyl sulfide, Bis(2-mercaptoethyl) sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HSCH2CH2)2S
CAS Number:
Molecular Weight:
154.32
Beilstein:
1445994
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.5961 (lit.)

bp

135-136 °C/10 mmHg (lit.)

density

1.183 g/mL at 25 °C (lit.)

SMILES string

SCCSCCS

InChI

1S/C4H10S3/c5-1-3-7-4-2-6/h5-6H,1-4H2

InChI key

KSJBMDCFYZKAFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zuoti Xie et al.
ACS nano, 11(1), 569-578 (2016-12-10)
The transport properties of molecular junctions based on alkanedithiols with three different methylene chain lengths were compared with junctions based on similar chains wherein every third -CH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service