Skip to Content
Merck
All Photos(1)

Key Documents

JKA8027

Sigma-Aldrich

8arm-PEG10K

average Mn 10,000, Maleimide, hexaglycerol core, thiol reactive

Synonym(s):

8arm-PEG-Maleimide, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
R(O(CH2CH2O)nCH2CH2NHCOCH2H2C4H2O2N)8
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

Product Name

8arm-PEG10K-Maleimide, hexaglycerol core, average Mn 10,000

form

solid

mol wt

average Mn 10,000

reaction suitability

reactivity: thiol reactive

polymer architecture

shape: 8-arm
functionality: homofunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

8arm PEG Maleimide. Crosslinks into hydrogels. MAL group is selective for thiol groups; reacts at pH 5.0-6.5.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu, H.D., et al.
Biomaterials, 33(7) (2012)

Articles

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service