Skip to Content
Merck
All Photos(1)

Key Documents

HAM203270

Hamilton® SaltLine reagent syringe

model, 1010 TLL-SAL, no needle, volume 10 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42142600
NACRES:
NB.03
Pricing and availability is not currently available.

material

Gastight syringe

feature

autoclavable: no
model 1010 TLL-SAL, no needle

manufacturer/tradename

Hamilton

parameter

20-50 °C temp. range (68-122 °F)
200 psig pressure

volume

10 mL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The SaltLine syringes are utilized in applications with high salt concentrations. They have the same PTFE Luer Lock termination but the plunger tip is made from Ultra High Molecular Weight Poly Ethylene (UHMWPE). The UHMWPE plunger tip resists damage that can be caused when salt crystal precipitate from the salt solution over time.

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the inner diameter of product HAM203270?

    1 answer
    1. The inner diameter of product HAM203270 is 14.6mm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service