Skip to Content
Merck
All Photos(1)

Key Documents

F6437

Sigma-Aldrich

Fmoc-(R)-2-(pentenyl)Ala-OH

≥97% (HPLC)

Synonym(s):

(R)-N-Fmoc-α-4-n-pentylalanine, Fmoc-(R)-2-(4-pentenyl)alanine, Fmoc-(R)-2-amino-2-methyl-hept-6-enoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25NO4
CAS Number:
Molecular Weight:
379.45
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

−20°C

SMILES string

O=C(OCC1C(C=CC=C2)=C2C3=C1C=CC=C3)N[C@@](CCCC=C)(C)C(O)=O

InChI

1S/C23H25NO4/c1-3-4-9-14-23(2,21(25)26)24-22(27)28-15-20-18-12-7-5-10-16(18)17-11-6-8-13-19(17)20/h3,5-8,10-13,20H,1,4,9,14-15H2,2H3,(H,24,27)(H,25,26)/t23-/m1/s1

InChI key

MRJFPZWLOJOINV-HSZRJFAPSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service