E45414
Ethyl 2-picolinate
99%
Synonym(s):
Ethyl pyridine-2-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
Beilstein:
122485
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
liquid
refractive index
n20/D 1.511 (lit.)
bp
240-241 °C (lit.)
mp
2 °C (lit.)
density
1.119 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)c1ccccn1
InChI
1S/C8H9NO2/c1-2-11-8(10)7-5-3-4-6-9-7/h3-6H,2H2,1H3
InChI key
FQYYIPZPELSLDK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
224.6 °F - closed cup
Flash Point(C)
107 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Arnau Garau et al.
Talanta, 133, 120-126 (2014-12-02)
A simple and expedient flow-based assembly capitalizing on programmable flow is herein proposed for reliable determination of trace level concentrations of aluminum as a potential contaminant in dialysis concentrate solutions without any prior sample clean-up/preconcentration procedure. Using salicylaldehyde picolinoylhydrazone in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service