Skip to Content
Merck
All Photos(2)

Key Documents

D75807

Sigma-Aldrich

2,5-Dichlorothiophene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2Cl2S
CAS Number:
Molecular Weight:
153.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.562 (lit.)

bp

162 °C (lit.)

density

1.442 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Clc1ccc(Cl)s1

InChI

1S/C4H2Cl2S/c5-3-1-2-4(6)7-3/h1-2H

InChI key

FGYBDASKYMSNCX-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service