All Photos(1)
About This Item
Linear Formula:
(CH3)2C6H2-1,4-(=O)2
CAS Number:
Molecular Weight:
136.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
solid
mp
71-73 °C (lit.)
SMILES string
CC1=CC(=O)C=C(C)C1=O
InChI
1S/C8H8O2/c1-5-3-7(9)4-6(2)8(5)10/h3-4H,1-2H3
InChI key
SENUUPBBLQWHMF-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A C Stewart
The Biochemical journal, 204(3), 705-712 (1982-06-15)
1. Photosynthetic electron transport from water to lipophilic Photosystem II acceptors was stimulated 3--5-fold by high concentrations (greater than or equal to 1 M) of salts containing anions such as citrate, succinate and phosphate that are high in the Hofmeister
Sam Hay et al.
The journal of physical chemistry. B, 111(13), 3488-3495 (2007-03-29)
The electrochemistry of 2,6-dimethylbenzoquinone (DMBQ) has been characterized for three different systems: DMBQ freely solvated in aqueous buffer; DMBQ bound to a neutral, blocked cysteine (N-acetyl-L-cysteine methyl ester) and the resulting DMBQ-bCys compound solvated in aqueous buffer; and DMBQ bound
D Kim et al.
Chemistry & biology, 5(2), 103-117 (1998-03-13)
The temporal and spatial control of the transition from vegetative to parasitic growth is critical to any parasite, but is essential to the sessile parasitic plants. It has been proposed that this transition in Striga spp. is controlled simply by
Mustafa Odabaşoğlu et al.
Acta crystallographica. Section C, Crystal structure communications, 61(Pt 4), o240-o242 (2005-04-05)
Molecules of the title compound, C8H9NO2, are linked into sheets by a combination of C-H...N, O-H...N and O-H...O hydrogen bonds and C-H...pi interactions. The hydrogen bonds are arranged as described by the graph-set ring notations R(2)(2)(7) and R(3)(3)(5), and a
Argyrides Argyrou et al.
Biochemistry, 42(7), 2218-2228 (2003-02-20)
Lipoamide dehydrogenase catalyzes the reversible NAD(+)-dependent oxidation of the dihydrolipoyl cofactors that are covalently attached to the acyltransferase components of the pyruvate dehydrogenase, alpha-ketoglutarate dehydrogenase, and glycine reductase multienzyme complexes. It contains two redox centers: a tightly, but noncovalently, bound
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service