CLS795225
Pyrex® Lifetime Red™ graduated tube
O.D. × L 25 mm × 200 mm, volume 50.0 mL, accuracy: 0.4 mL
Synonym(s):
Corning® 7952
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
round bottom tube
packaging
case of 1
pack of 1
manufacturer/tradename
Corning 7952-25
volume measuring range
50.0 mL, accuracy: 0.4 mL
O.D. × L
25 mm × 200 mm
Related Categories
General description
Used for general hospital laboratory work. It was designed by Dr. F.N.L. Taylor, Boston City Hospital, for determinations involving multiple calibrations and for carrying out digestions. Graduated at 12.5, 25, 35, and 50 mL, and supplied with a durable, white enamel marking spot.
Legal Information
Corning is a registered trademark of Corning, Inc.
Lifetime-Red is a trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Prehospital and disaster medicine, 35(5), 482-487 (2020-07-01)
To date, all human studies of mass-casualty decontamination for chemical incidents have relied on the collection and analysis of external samples, including skin and hair, to determine decontamination efficacy. The removal of a simulant contaminant from the surface of the
Experimental animals, 69(4), 423-429 (2020-06-20)
The cold storage of two-cell embryos is a useful technique for transporting genetically engineered mice without the shipment of live animals. However, the developmental ability of cold-stored embryos decreases with prolonged storage periods. Therefore, the transported embryos must be readily
Scientific reports, 10(1), 609-609 (2020-01-19)
Since the first work by Laurencin and colleagues on the development of polymeric electrospinning for biomedical purposes, the use of electrospinning technology has found broad applications in such areas of tissue regeneration and drug delivery. More recently, coaxial electrospinning has
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 36(12), 1787-1799 (2019-09-26)
Iodised salt (supplemented with potassium iodide) is the primary source of iodine in Switzerland, but it is rarely used in the manufacture of cheese. In the present study, the diffusion of iodide and chloride in experimentally produced soft, semi-hard and
Animal reproduction science, 210, 106197-106197 (2019-10-23)
Cryopreservation in liquid nitrogen (LN2) allows for semen to be stored for long periods of time while there is sustaining of sperm viability. In this study, there was assessment of effects induced by different storage temperatures on cryopreserved dog spermatozoa.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service