Skip to Content
Merck
All Photos(1)

Key Documents

CLS44202XL

Pyrex® Fernbach culture flask

capacity 2,800 mL

Synonym(s):

Corning® 4420

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.43
Pricing and availability is not currently available.

material

borosilicate glass
flat bottom flask
wide-mouth
wide-mouth neck

Joint

joint stopper13

sterility

non-sterile

feature

cap: no
closure type Beaded rim

packaging

case of 6

manufacturer/tradename

Corning 4420-2XL

O.D. × H

~205 mm × 230 mm

capacity

2,800 mL

Looking for similar products? Visit Product Comparison Guide

General description

Wide-mouth design for culturing organisms requiring a large surface area to volume ratio. Can also be used in serum production. Reference: Industrial and Engineering Chemistry, Vol. 21, No. 12, 1198.
Note: Do not use this flask on a heat source smaller than the base of the flask. Uneven heating of the flask could cause breakage.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service