Skip to Content
Merck
All Photos(1)

Key Documents

CDS000677

Sigma-Aldrich

1-Phenylethyl mercaptan

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10S
Molecular Weight:
138.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

liquid

SMILES string

CC(S)c1ccccc1

InChI

1S/C8H10S/c1-7(9)8-5-3-2-4-6-8/h2-7,9H,1H3

InChI key

QZZBJCFNHPYNKO-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Nervous system, Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Schoenauer et al.
Journal of agricultural and food chemistry, 67(9), 2598-2606 (2019-02-15)
Following a structure/odor activity approach as previously published, the present study was focused on three aromatic thiols also identified as food odorants, namely 1-phenylethane-1-thiol, phenyl methanethiol, and 2-phenylethanethiol. Their structures were systematically modified to receive 16 new sulfur-containing benzene derivatives.
Korath Shivan Sugi et al.
The Analyst, 145(4), 1337-1345 (2020-01-22)
The synthesis of atomically precise noble metal clusters using various protocols often results in a mixture of clusters with different cores. Hence, it is important to isolate such clusters in their pure form in terms of composition especially for crystallization.
Xiangming Meng et al.
Nanoscale research letters, 7(1), 277-277 (2012-06-01)
This work presents a controlled reduction method for the selective synthesis of different sized gold nanoclusters protected by thiolate (SR = SC2H4Ph). Starting with Au(III) salt, all the syntheses of Aun(SR)m nanoclusters with (n, m) = (20, 16), (24, 20), (39, 29), and (40
Goonay Yousefalizadeh et al.
Nanoscale, 13(1), 242-252 (2020-12-18)
Noble metal clusters have unique photophysical properties, especially as a new class of materials for multiphoton biomedical imaging. The previously studied Au25SR18 exhibits "giant" two-photon absorbance cross sections. Herein, we investigate the origins of the large two photon absorption for
Ananya Baksi et al.
Analytical chemistry, 90(19), 11351-11357 (2018-09-02)
A detailed mass-spectrometric study of atomically precise monolayer-protected clusters revealed the potential application of such materials as mass-spectrometric standards, mostly in negative-ion mode and in the high-mass range. To date, very few molecules are known that can be efficiently ionized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service