Skip to Content
Merck
All Photos(1)

Key Documents

C95609

Sigma-Aldrich

Cyclobutanecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H7CO2H
CAS Number:
Molecular Weight:
100.12
Beilstein:
1816777
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

liquid

refractive index

n20/D 1.444 (lit.)

bp

195 °C (lit.)

mp

−7.5 °C (lit.)

density

1.047 g/mL at 25 °C (lit.)

SMILES string

OC(=O)C1CCC1

InChI

1S/C5H8O2/c6-5(7)4-2-1-3-4/h4H,1-3H2,(H,6,7)

InChI key

TXWOGHSRPAYOML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian D Hudson et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 26(12), 4951-4965 (2012-08-25)
When it is difficult to develop selective ligands within a family of related G-protein-coupled receptors (GPCRs), chemically engineered receptors activated solely by synthetic ligands (RASSLs) are useful alternatives for probing receptor function. In the present work, we explored whether a
Paolo Lorenzon et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(1), 146-160 (2015-01-09)
The type of neuronal activity required for circuit development is a matter of significant debate. We addressed this issue by analyzing the topographic organization of the olfactory bulb in transgenic mice engineered to have very little afferent spontaneous activity due

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service