Skip to Content
Merck
All Photos(2)

Key Documents

BR781354

BRAND® 96-well PCR plate

non-skirted, white, cut corner H12, standard profile

Synonym(s):

PCR, PCR Plates, Plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
material:
white
white polypropylene
size:
96 well
binding type:
non-treated surface
feature:
skirt: no
No skirt
blue coding
standard profile
Pricing and availability is not currently available.

material

white
white polypropylene

description

D-Nase/R-Nase, DNA-free, suitable for qPCR

feature

skirt: no
No skirt
blue coding
standard profile

packaging

pack of 50 ea

manufacturer/tradename

BRAND 781354

technique(s)

PCR: suitable

size

96 well

working volume

200 μm

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

PCR plates consisting of 96 PCR tubes (0.2 mL), with alphanumeric coding. Ultra-thin wall design facilitates rapid and steady heat transfer. Smooth interior tube surface minimizes binding of enzymes and nucleic acids. Raised rims of wells protect against cross-contamination and provide tight seal particularly in combination with BRAND´s new sealing mats. Furthermore these sealing mats can be easily pierced with standard pipette tips. The PCR-plates can also be sealed with traditional self-adhesive sealing films. Skirted plates are available with bar code on request.

Features and Benefits

  • BRAND® PCR plates consisting of 96 PCR tubes (0.2 mL), with alphanumeric coding.
  • Ultra-thin wall design facilitates rapid and steady heat transfer.
  • Smooth interior tube surface minimizes binding of enzymes and nucleic acids.
  • Raised rims of wells protect against cross-contamination and provide tight seal particularly in combination with BRAND´s new sealing mats.
  • Furthermore these sealing mats can be easily pierced with standard pipette tips.
  • The PCR-plates can also be sealed with traditional self-adhesive sealing films.
  • Skirted plates are available with bar code on request.
  • Suitable for all standard thermal cyclers.
  • Compatible with cap strips and sealing films to protect against evaporation and contamination.
  • Ideal for PCR and qPCR applications.
  • Fully-skirted plates, are ideal for automation due to their rigidity.
  • Unskirted plates for high flexibility.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service