BR781039
BRAND® PC centrifuge tube, rimless
volume 100 mL, round bottom, L × W 41 mm × 115 mm
Synonym(s):
Centrifuge tubes, Tubes
About This Item
Recommended Products
material
polycarbonate
round bottom
transparent tube
description
RCF up to 20000g
feature
RCF 20,000 × g
packaging
pack of 10
manufacturer/tradename
BRAND 781039
L × W
41 mm × 115 mm
volume
100 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service