BR702886
BRAND® combi-pack caps and seal for Transferpettor
2000-10000 μL, 2 PP caps, 1 PE seal
Synonym(s):
702886, Accessories, Caps, Pipettes, Piston-operated pipettor, Positive displacement pipettor, Seals, Transferpettor
About This Item
Recommended Products
material
polyethylene seal
polypropylene cap
sterility
non-sterile
packaging
pack of 1 set
manufacturer/tradename
BRAND 702886
maximum volume
10 mL
Looking for similar products? Visit Product Comparison Guide
General description
The Transferpettor operates on the positive displacement principle. In contrast with air displacement pipettes, the piston is in direct contact with the aspirated liquid. The tightly fitting piston always wipes the walls of the tips completely clean - literally to the last drop. Therefore providing reproducible results. There is no need to discard tips after each pipetting operation, since residual wetting is negligible.
Features and Benefits
- Density up to 3.8 g/cm
- Vapor pressure up to 500 mbar
- Kinematic viscosity up to: Nominal volume 1/ 2/ 5 µl: 6.000 mm² /s; nominal volume 10 µl: 20.000 mm² /s; nominal volume 25 µl: 50.000 mm² /s; nominal volume 50 µl: 100.000 mm² /s; nominal volume 100 µl: 140.000 mm² /s (Operating temperature: 15 - 40 °C (59 °F - 104 °F))
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service