Key Documents
BR29125
BRAND® pipette rinsing system, rinser for pipette
high-density polyethylene, capacity 17 L, L × I.D. 1000 mm × 150 mm
Synonym(s):
Pipette Rinsing Systems, Pipette rinser, Pipettes
About This Item
UNSPSC Code:
41103200
NACRES:
NB.35
material
high-density polyethylene
manufacturer/tradename
BRAND 29125
L × I.D.
1000 mm × 150 mm
capacity
17 L
Related Categories
General description
PE-HD. Trouble-free and gentle cleaning of pipettes with repetitive siphon rinsing. Water supply connection with hose nozzle outer Ø 12 mm. Soaking jar and pipette basket sold separately. Adjust the water flow to approx. 2 L/min.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)