Key Documents
BR152510
BRAND® tubing adapter
high-density polyethylene, for tubing i.d., 12-14 to 18-20,5 mm
Synonym(s):
Tubing, Tubing adapters
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
material
high-density polyethylene
feature
for tubing i.d. 12-14 to 18-20,5 mm
packaging
pack of 10 ea
manufacturer/tradename
BRAND 152510
L
73 mm
Related Categories
General description
The BRAND® tubing adapter is designed to provide a secure connection. Its translucent construction allows for easy visibility of fluid flow, facilitating monitoring during use.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)