Key Documents
BMSV0020
Sigma-Aldrich
Aspire Laboratory Aspirator
includes base with internal vacuum pump, 2 L Polycarbonate bottle with lid, silicone tubing, handheld vacuum controller and single channel adapters, AC/DC input 115 V AC, US 3-pin plug
Synonym(s):
aspiration system, vacuum aspirator
About This Item
UNSPSC Code:
41161502
Pricing and availability is not currently available.
manufacturer/tradename
Benchmark Scientific V0020
parameter
-0 mbar vacuum (-750 mbar)
outside W × D × H
9.5 in. × 7.9 in. × 13.9 in.
General description
The ASPIRE Laboratory Aspirator incorporates a quiet, maintenance-free pump inside a sturdy, yet compact base. The vacuum is fully adjustable, with a pressure gauge that displays the vacuum level, allowing full control of aspirating speed. The ASPIRE is an ideal choice for a wide range of laboratory aspirating applications, including removal of media from tissue culture vessels or plates, and supernatants.
Legal Information
Aspire is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)