Skip to Content
Merck
All Photos(1)

Key Documents

B10587

Sigma-Aldrich

1-Benzosuberone

99%

Synonym(s):

6,7,8,9-Tetrahydro-5-benzocycloheptenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O
CAS Number:
Molecular Weight:
160.21
Beilstein:
636823
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.564 (lit.)

bp

270 °C (lit.)

density

1.071 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCc2ccccc12

InChI

1S/C11H12O/c12-11-8-4-2-6-9-5-1-3-7-10(9)11/h1,3,5,7H,2,4,6,8H2

InChI key

KWHUHTFXMNQHAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service