ALD00613
PSI2
Synonym(s):
Ψ2, 2-(Perfluorophenoxy)-1,3,2-dithiaphospholane 2-sulfide, Achiral phosphorus-sulfur Incorporation reagent
About This Item
Recommended Products
Application
Learn more about PI and PSI reagents
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
PI and PSI reagents are a platform of phosphate-based reagents for phosphorylation and phosphothiolation of nucleotides and peptides developed by the Baran lab.
PI and PSI reagents are a platform of phosphate-based reagents for phosphorylation and phosphothiolation of nucleotides and peptides developed by the Baran lab.
PI and PSI reagents are a platform of phosphate-based reagents for phosphorylation and phosphothiolation of nucleotides and peptides developed by the Baran lab.
PI and PSI reagents are a platform of phosphate-based reagents for phosphorylation and phosphothiolation of nucleotides and peptides developed by the Baran lab.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service