Skip to Content
Merck
All Photos(1)

Key Documents

934267

Sigma-Aldrich

Ready-to-Cast Lithium Titanate (Li4Ti5O12) Slurry for Lithium ion battery

Blade coatable ink slurry for Lithium ion battery

Synonym(s):

LTO anode slurry

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Li4O12Ti5
Molecular Weight:
459.09
UNSPSC Code:
12352402
NACRES:
NA.21

grade

battery grade

Quality Level

form

slurry

composition

Active material, >15 wt. %
Solid content, 22-27 wt. %

particle size

1-3 μm

application(s)

battery manufacturing

General description

Our high-quality ready-to-cast lithium titanate (LTO) anode slurry is a liquid dispersion consisting of LTO active material, conductive additives, binder, and organic solvent. Its low moisture content allows for the production of stable anode films with extended cycle life.

Application

Our LTO anode slurry is formulated for use in secondary lithium-ion batteries and can be directly applied using blade-coating or slot-die coating onto copper foil current collectors. The stable and easy processing properties of our product promise high-quality, defect-free LTO anodes with excellent performance.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

195.8 °F

Flash Point(C)

91 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conveying Advanced Li-ion Battery Materials into Practice The Impact of Electrode Slurry Preparation Skills
Kraytsberg A, et al.
Advanced Energy Materials, 6, 1600655-1600655 (2016)
An Effective Mixing for Lithium Ion Battery Slurries
Liu D, et al.
Advances in Chemical Engineering and Science, 4, 515 - 528 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service