Skip to Content
Merck
All Photos(1)

Key Documents

930970

Sigma-Aldrich

Magnesium chloride hexahydrate

99.9% trace metals basis

Synonym(s):

Magnesium dichloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
grade:
for analytical purposes
form:
powder, crystals or chunks
solubility:
acetone: slightly soluble
alcohols: slightly soluble
water: soluble (highly soluble)
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

Assay

99.9% trace metals basis

form

powder, crystals or chunks

mp

117 °C

solubility

acetone: slightly soluble
alcohols: slightly soluble
water: soluble (highly soluble)

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium chloride hexahydrate is a colorless salt. The salt is soluble in water and slightly soluble in ethanol, acetone, and pyridine. Chemically, magnesium chloride is a weak Lewis acid.
Magnesium chloride is extracted from naturally occurring brine, for example by solar evaporation of sea water, and purified by solution chemistries.

Application

Magnesium chloride is used as a source of the divalent magnesium ion, often to control zeta potential and ionic strength. Commonly, magnesium chloride is used in water desalination experiments, particularly research investigating nanofiltration membranes. In addition, researchers have used magnesium chloride to alter the stability of colloidal suspensions, for example driving the coagulation of graphene oxide.[1] Magnesium chloride has also been used to control the interlayer spacing of graphene-oxide membranes[2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ion sieving in graphene oxide membranes via cationic control of interlayer spacing.
Chen, L., et al.
Bio/technology (Nature Publishing Company), 550, 380?383-380?383 (2017)
Aggregation and Stability of Reduced Graphene Oxide: Complex Roles of Divalent Cations, pH, and Natural Organic Matter
Chowdhury, I., et al.
Environmental Science & Technology, 49, 10886-10893 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service