Skip to Content
Merck
All Photos(2)

Key Documents

914584

Sigma-Aldrich

Phenylalanine-4′-azobenzene HCl

≥95%

Synonym(s):

(2S)-2-Amino-3-[4-(2-phenyldiazen-1-yl)phenyl]propanoic acid hydrochloride, (E)-2-Amino-3-(4-(phenyldiazenyl)phenyl)propanoic acid HCl, 4-(2-Phenyldiazenyl)-L-phenylalanine hydrochloride, 4-(Phenylazo)-L-phenylalanine, AzoF, Photoswitchable phenylalanine amino acid, Reversible photo-controllable unnatural amino acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16ClN3O2
CAS Number:
Molecular Weight:
305.76
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

storage temp.

2-8°C

SMILES string

Cl.N[C@@H](Cc1ccc(cc1)\N=N\c2ccccc2)C(=O)O

InChI key

LRSKKAMIJGFIDZ-GUFYRWSKSA-N

Application

Phenylalanine-4′-azobenzene HCl (AzoF) is a phenylalanine derivative with an azo bond that isomerizes upon UV irradiation from its E isomer to its Z isomer and reverts back at light above 400 nm. When incorporated into proteins or chemical probes, this reversibility is utilized as a photoswitch with spatial and temporal regulation within biological systems.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea C Kneuttinger et al.
Cell chemical biology, 26(11), 1501-1514 (2019-09-10)
Imidazole glycerol phosphate synthase (ImGPS) is an allosteric bienzyme complex in which substrate binding to the synthase subunit HisF stimulates the glutaminase subunit HisH. To control this stimulation with light, we have incorporated the photo-responsive unnatural amino acids phenylalanine-4'-azobenzene (AzoF)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service