Skip to Content
Merck
All Photos(2)

Key Documents

912476

Sigma-Aldrich

(S)-AntPhos

≥97%

Synonym(s):

(S)-4-(Anthracen-9-yl)-3-(tert-butyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H23OP
CAS Number:
Molecular Weight:
370.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Quality Level

Assay

≥97%

form

powder

optical purity

ee: ≥99% (HPLC)

reaction suitability

reagent type: ligand

functional group

phosphine

SMILES string

P1(COc2c1c(ccc2)c3c4c(cc5c3cccc5)cccc4)C(C)(C)C

InChI

1S/C25H23OP/c1-25(2,3)27-16-26-22-14-8-13-21(24(22)27)23-19-11-6-4-9-17(19)15-18-10-5-7-12-20(18)23/h4-15H,16H2,1-3H3

InChI key

HKAWVLHXUZNLPP-UHFFFAOYSA-N

Application

(S)-AntPhos is a P-chiral monophosphorus ligand used for the asymmetric Suzuki-Miyaura and Miyaura borylation reactions. This ligand is uniquely effective for sterically hindered cross-coupling reactions.

Legal Information

Sold in collaboration with Zejun Pharmaceuticals

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenzhen Fu et al.
Angewandte Chemie (International ed. in English), 54(8), 2520-2524 (2015-01-20)
The first asymmetric nickel-catalyzed intramolecular reductive cyclization of alkynones is reported. A P-chiral monophosphine and triethylsilane were used as the ligand and the reducing reagent, respectively, to form a series of tertiary allylic alcohols bearing furan/pyran rings in excellent yields
Ruofei Cheng et al.
Journal of the American Chemical Society, 140(13), 4508-4511 (2018-03-27)
Carborane cage chirality is an outstanding issue of great interest as the icosahedral carboranes have wide applications in medicinal and materials chemistry. The synthesis of optically active carborane derivatives, whose chirality is associated with the substitution patterns on the polyhedron
Naifu Hu et al.
Journal of the American Chemical Society, 137(21), 6746-6749 (2015-05-06)
The rhodium-catalyzed asymmetric hydroboration of α-arylenamides with BI-DIME as the chiral ligand and (Bpin)2 as the reagent yields for the first time a series of α-amino tertiary boronic esters in good yields and excellent enantioselectivities (up to 99% ee).
Naifu Hu et al.
Angewandte Chemie (International ed. in English), 55(16), 5044-5048 (2016-03-19)
A highly enantioselective alkene aryloxyarylation led to the high-yielding formation of a series of 1,4-benzodioxanes, 1,4-benzooxazines, and chromans containing quaternary stereocenters with excellent enantioselectivity. The sterically bulky and conformationally well defined chiral monophosphorus ligand L4 or L5 was responsible for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service