Skip to Content
Merck
All Photos(2)

Key Documents

905097

Sigma-Aldrich

Ir(p-Me-ppy)3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H30IrN3
CAS Number:
Molecular Weight:
696.86
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

Looking for similar products? Visit Product Comparison Guide

Application

This versatile photocatalyst facilitates a variety of transformations using visible light.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Organofluorine chemistry is an essential part of drug discovery programs as well as agrochemical programs and even plays a major role in materials chemistry. Despite the undeniable importance of fluorinated organic molecules, our ability to synthesize these substrates is lacking - though arguably it is better than that of Nature. Consequently, methods that allow facile access to fluorinated molecules are important especially when they provide unique access to fluorinated chemical space.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service