Skip to Content
Merck
All Photos(1)

Key Documents

902578

Sigma-Aldrich

Hydroxy-PEG6-t-butyl ester

≥95%

Synonym(s):

tert-Butyl 1-hydroxy-3,6,9,12,15,18-hexaoxahenicosan-21-oate, HO-PEG6-CO-OtBu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H38O9
CAS Number:
Molecular Weight:
410.50
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥95%

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n/D 1.454

density

1.068

functional group

ester
hydroxyl

storage temp.

−20°C

SMILES string

O=C(OC(C)(C)C)CCOCCOCCOCCOCCOCCOCCO

InChI

1S/C19H38O9/c1-19(2,3)28-18(21)4-6-22-8-10-24-12-14-26-16-17-27-15-13-25-11-9-23-7-5-20/h20H,4-17H2,1-3H3

InChI key

VGGDPFAYSOSIOK-UHFFFAOYSA-N

Application

This heterobifunctional, PEGylated crosslinker features a hydroxyl group at one end and t-butyl-protected carboxylic acid at the other, which can be deprotected with acidic conditions. The hydrophillic PEG linker facilitates solubility in biological applications. Hydroxy-PEG6-t-butyl ester can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or roteolysis-targeting chimeras (PROTAC® molecules) for targeted protein degradation.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yury E Tsvetkov et al.
Journal of the American Chemical Society, 134(1), 426-435 (2011-11-18)
The human natural killer cell carbohydrate, HNK-1, plays function-conducive roles in peripheral nerve regeneration and synaptic plasticity. It is also the target of autoantibodies in polyneuropathies. It is thus important to synthesize structurally related HNK-1 carbohydrates for optimizing its function-conducive
André Warnecke et al.
Bioconjugate chemistry, 14(2), 377-387 (2003-03-20)
In situ binding of thiol-reactive prodrugs to the cysteine-34 position of circulating albumin is a new approach in drug delivery. Therefore, five maleimide-bearing derivatives of the anticancer drug camptothecin (CPT) were developed as albumin-binding prodrugs. These compounds were synthesized by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service