Skip to Content
Merck
All Photos(1)

Key Documents

900585

Sigma-Aldrich

(R)-2-Methylpiperidine

95%

Synonym(s):

(-)-2-Methylpiperidine, (-)-Pipecoline, (2R)-2-Methylpiperidine, (R)-2-Methylcyclohexylamine, 2-(R)-Methylpiperidine, R-(-)-2-Methylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N
CAS Number:
Molecular Weight:
99.17
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

liquid

format

neat

storage temp.

2-8°C

SMILES string

N1[C@@H](CCCC1)C

InChI

1S/C6H13N/c1-6-4-2-3-5-7-6/h6-7H,2-5H2,1H3/t6-/m1/s1

InChI key

NNWUEBIEOFQMSS-ZCFIWIBFSA-N

Application

Chiral building block developed using Liverpool ChiroChem-patented technology.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

58.0 °F

Flash Point(C)

14.44 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service