Skip to Content
Merck
All Photos(1)

Key Documents

900399

Sigma-Aldrich

SLAP N-Bn Pip Reagent

95%

Synonym(s):

N1-Benzyl-N1-((trimethylsilyl)methyl)ethane-1,2-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24N2Si
CAS Number:
Molecular Weight:
236.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid (paste)

refractive index

n/D 1.5071

density

0.9366 g/mL

functional group

amine
phenyl

storage temp.

−20°C

SMILES string

[Si](CN(CCN)Cc1ccccc1)(C)(C)C

InChI

1S/C13H24N2Si/c1-16(2,3)12-15(10-9-14)11-13-7-5-4-6-8-13/h4-8H,9-12,14H2,1-3H3

Application

Silicon amine protocol (SLAP) reagents, in conjunction with aldehydes and ketones, form N-unprotected piperazines via photocatalytic cross-coupling with Ir[(ppy)2dtbbpy]PF6 (747769). Heteroaromatic, aromatic, and aliphatic aldehydes are well tolerated, as are structurally and stereochemically complex SLAP reagents. This product was introduced in collaboration with the Bode Research Group and provides a tin-free alternative to SnAP (tin amine protocol) reagents for piperazine synthesis.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

198.0 °F

Flash Point(C)

92.2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng-Ying Hsieh et al.
ACS central science, 3(1), 66-72 (2017-02-06)
Redox neutral photocatalytic transformations often require careful pairing of the substrates and photoredox catalysts in order to achieve a catalytic cycle. This can limit the range of viable transformations, as we recently observed in attempting to extend the scope of
Sheng-Ying Hsieh et al.
Organic letters, 18(9), 2098-2101 (2016-04-22)
Silicon amine protocol (SLAP) reagents for photocatalytic cross-coupling with aldehydes and ketones to form N-unprotected piperazines have been developed. This blue light promoted process tolerates a wide range of heteroaromatic, aromatic, and aliphatic aldehydes and structurally and stereochemically complex SLAP
Moritz K Jackl et al.
Organic letters, 19(17), 4696-4699 (2017-08-17)
Photocatalytic coupling of aldehydes and silicon amine protocol (SLAP) reagents enables the simple, scalable synthesis of substituted morpholines, oxazepanes, thiomorpholines, and thiazepanes under continuous flow conditions. Key to the success of this process is the combination of an inexpensive organic

Protocols

The expanding class of SLAP reagents provides access to saturated N-heterocycles and aliphatic aldehydes and ketones without additional reagents or the generation of toxic byproducts.

The expanding class of SLAP reagents provides access to saturated N-heterocycles and aliphatic aldehydes and ketones without additional reagents or the generation of toxic byproducts.

The expanding class of SLAP reagents provides access to saturated N-heterocycles and aliphatic aldehydes and ketones without additional reagents or the generation of toxic byproducts.

The expanding class of SLAP reagents provides access to saturated N-heterocycles and aliphatic aldehydes and ketones without additional reagents or the generation of toxic byproducts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service