Skip to Content
Merck
All Photos(1)

Key Documents

87921

Sigma-Aldrich

Tetramethylsilane

≥99.0% (GC)

Synonym(s):

TMS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si(CH3)4
CAS Number:
Molecular Weight:
88.22
Beilstein:
1696908
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥99.0% (GC)
bp:
26-28 °C (lit.)
vapor pressure:
11.66 psi ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

11.66 psi ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

842 °F

refractive index

n20/D 1.358 (lit.)
n20/D 1.359

bp

26-28 °C (lit.)

mp

−99 °C (lit.)

density

0.648 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)C

InChI

1S/C4H12Si/c1-5(2,3)4/h1-4H3

InChI key

CZDYPVPMEAXLPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetramethylsilane can be used as a silicon precursor for the synthesis of silicon doped diamond-like carbon (DLC-Si) films[1][2] and silicon carbide (SiC) bulk crystals.[3] It can also be used as a hydrocarbon substrate to study intermolecular C-H activation chemistry.[4]

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-16.6 °F - closed cup

Flash Point(C)

-27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The mechanical and biocompatibility properties of DLC-Si films prepared by pulsed DC plasma activated chemical vapor deposition.
Bendavid A, et al.
Diamond and Related Materials, 16(8), 1616-1622 (2007)
High-temperature chemical vapor deposition for SiC single crystal bulk growth using tetramethylsilane as a precursor.
Nam DH, et al.
Crystal Growth & Design, 14(11), 5569-5574 (2014)
The effects of Si incorporation on the electrochemical and nanomechanical properties of DLC thin films.
Papakonstantinou P, et al.
Diamond and Related Materials, 11(3-6), 1074-1080 (2002)
Intermolecular C- H Activation of Hydrocarbons by Tungsten Alkylidene Complexes: An Experimental and Computational Mechanistic Study.
Adams CS, et al.
Organometallics, 20(23), 4939-4955 (2001)
Jose C Pichardo et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 52(9), 1482-1489 (2011-07-30)
The current gold standard for measuring marrow cellularity is the bone marrow (BM) biopsy of the iliac crest. This measure is not predictive of total marrow cellularity, because the biopsy volume is typically small and fat fraction varies across the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service