Skip to Content
Merck
All Photos(3)

Key Documents

856088

Sigma-Aldrich

β-Cyclodextrin hydrate

99%

Synonym(s):

beta-Cyclodextrin hydrate, β-Schardinger dextrin, Cycloheptaamylose, Schardinger β-Dextrin hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H70O35 · xH2O
CAS Number:
Molecular Weight:
1134.98 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

solid

optical activity

[α]20/D +142°, c = 1 in H2O (Dry basis)

mp

>260 °C (dec.) (lit.)

functional group

ether

SMILES string

O.OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C42H70O35.H2O/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51;/h8-63H,1-7H2;1H2/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-;/m1./s1

InChI key

JNSAKCOAFBFODP-ZQOBQRRWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-cyclodextrin (β-CD), a well-known macrocyclic oligosaccharide member of the CD family, is composed of seven D-glucose units connected via α(1→4) glycosidic bonds. Its ability to form several inclusion complexes with organic as well as inorganic compounds makes it a potential candidate in solid and solution state determinations.[1][2]

Application

β-Cyclodextrin hydrate may be used:
  • To form an inclusion complex with norfloxacin for subsequent analysis using high performance liquid chromatography (HPLC).[3]
  • As a precursor of heptakis-(2,3-diacetyl-6-sulfato)-β-cyclodextrin for use as a chiral selector in the enantioseparation of linezolid by capillary electrophoresis (CE).[4]

Forms clathrates. Produces a water-soluble complex with dansyl chloride (cat. no. D143359) for the fluorescent labeling of proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Michalska et al.
Journal of chromatography. A, 1180(1-2), 179-186 (2008-01-01)
A method for the enantioseparation of linezolid, the first compound of a truly new class of antibiotics-the oxazolidinones, was developed. The elaborated method of linezolid enantiomers separation was successfully performed using an anionic single-isomer cyclodextrin-heptakis-(2,3-diacetyl-6-sulfato)-beta-cyclodextrin (HDAS-beta-CD) as a resolving agent
R G Winkler et al.
Journal of computer-aided molecular design, 14(7), 659-667 (2000-09-29)
We study by molecular dynamics simulations the hydration of beta-cyclodextrin. Our simulations show that within these barrel-shaped molecules hydrophobicity dominates, while at the top and bottom sides of the barrel interactions with water are mostly hydrophilic in nature. These results
Cassiana Mendes et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(10), 1083-1089 (2015-10-16)
The aim of this study was to develop and validate a simple liquid-chromatography method, with good accuracy, reproducibility and sensitivity, for the quantification of norfloxacin in β-cyclodextrin inclusion complexes. In the method validation, the parameters evaluated were linearity, limits of
beta-Cyclodextrin hydration: a calorimetric and gravimetric study
Bilal M, et al.
Thermochimica Acta, 249, 63-73 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service