Skip to Content
Merck
All Photos(4)

Key Documents

771406

Sigma-Aldrich

Zinc trifluoromethanesulfinate

Synonym(s):

1,1,1-Trifluoro-methanesulfinic acid zinc salt (2:1), Baran trifluoromethylation reagent, Bis(((trifluoromethyl)sulfinyl)oxy)zinc, TFMS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2F6O4S2Zn
CAS Number:
Molecular Weight:
331.53
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22
Pricing and availability is not currently available.

form

solid

Quality Level

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reaction type: C-H Activation

mp

151-157 °C

storage temp.

2-8°C

SMILES string

FC(F)(F)S(=O)O[Zn]OS(=O)C(F)(F)F

InChI

1S/2CHF3O2S.Zn/c2*2-1(3,4)7(5)6;/h2*(H,5,6);/q;;+2/p-2

InChI key

UANOWFITUWBPCF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Zinc trifluoromethanesulfinate (TFMS) is part of a zinc sulfinate toolbox developed by Phil Baran for the simple and rapid diversification of heterocycles.[1]

Practical and Innate Carbon-Hydrogen Functionalization of Heterocycles

Learn More at the Professor and Product Portal of Professor Phil S. Baran.

Linkage

Frequently Asked Questions are available for this Product.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuta Fujiwara et al.
Nature, 492(7427), 95-99 (2012-12-04)
Nitrogen-rich heterocyclic compounds have had a profound effect on human health because these chemical motifs are found in a large number of drugs used to combat a broad range of diseases and pathophysiological conditions. Advances in transition-metal-mediated cross-coupling have simplified
Fionn O'Hara et al.
Journal of the American Chemical Society, 135(32), 12122-12134 (2013-07-19)
Radical addition processes can be ideally suited for the direct functionalization of heteroaromatic bases, yet these processes are only sparsely used due to the perception of poor or unreliable control of regiochemistry. A systematic investigation of factors affecting the regiochemistry

Articles

Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.

Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.

Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.

Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service