Skip to Content
Merck
All Photos(2)

Key Documents

748161

Sigma-Aldrich

Silica

Silica

nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm

Synonym(s):

Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

mesoporous
nanoparticles

Quality Level

particle size

200 nm

pore size

4 nm pore size

bp

2230 °C (lit.)

mp

>1600 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Shape - approximately spherical

Application

The well defined mesoporous materials are used in applications that include separations[1], drug delivery[2], optical imaging[3], and composites.[4]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thiam-Leng Chew et al.
Advances in colloid and interface science, 153(1-2), 43-57 (2010-01-12)
Separation of carbon dioxide (CO(2)) from gaseous mixture is an important issue for the removal of CO(2) in natural gas processing and power plants. The ordered mesoporous silicas (OMS) with uniform pore structure and high density of silanol groups, have
Zhang, D.; Wang, X.; Qiao, Z.; Tang, D.; Liu, Y.; Huo, Q.
The Journal of Physical Chemistry C, 114, 12505-12505 (2010)
Srivalleesha Mallidi et al.
Nano letters, 9(8), 2825-2831 (2009-07-04)
Gold nanoparticles targeting epidermal growth factor receptor via antibody conjugation undergo molecular specific aggregation when they bind to receptors on cell surfaces, leading to a red shift in their plasmon resonance frequency. Capitalizing on this effect, we demonstrate the efficacy
Huan Meng et al.
ACS nano, 4(8), 4539-4550 (2010-08-25)
Overexpression of drug efflux transporters such as P-glycoprotein (Pgp) protein is one of the major mechanisms for multiple drug resistance (MDR) in cancer cells. A new approach to overcome MDR is to use a co-delivery strategy that utilizes a siRNA
Wang, Z.; Stein, A.
Chemistry of Materials, 20, 1029-1029 (2008)

Articles

RAFT polymerization controls chain growth, yields defined polymers without cytotoxic heavy metals like ATRP.

RAFT polymerization controls chain growth, yields defined polymers without cytotoxic heavy metals like ATRP.

RAFT polymerization controls chain growth, yields defined polymers without cytotoxic heavy metals like ATRP.

RAFT polymerization controls chain growth, yields defined polymers without cytotoxic heavy metals like ATRP.

See All

Questions

1–3 of 3 Questions  
  1. Is this silica nanoparticle hydrophobic?

    1 answer
    1. This product does not have a contact angle measurement to test its hydrophobicity. Generally, non-functionalized silica is hydrophilic.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service