Skip to Content
Merck
All Photos(1)

Key Documents

739782

Sigma-Aldrich

5-Methyl-2-(tributylstannyl)pyridine

95%

Synonym(s):

Tributyl-(5-methylpyridin-2-yl)stannane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H33NSn
CAS Number:
Molecular Weight:
382.17
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
95%

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.514

density

1.106 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1ccc(C)cn1

InChI

1S/C6H6N.3C4H9.Sn/c1-6-3-2-4-7-5-6;3*1-3-4-2;/h2-3,5H,1H3;3*1,3-4H2,2H3;

InChI key

MVOHAZAWWAXIDR-UHFFFAOYSA-N

Application

Used in Stille cross coupling reactions.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service