Skip to Content
Merck
All Photos(1)

Key Documents

734462

Sigma-Aldrich

Methyl 2-(chloromethyl)acrylate

contains ≤0.5% BHT as inhibitor, 95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7ClO2
CAS Number:
Molecular Weight:
134.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

liquid

contains

≤0.5% BHT as inhibitor

refractive index

n20/D 1.455

density

1.150 g/mL at 25 °C

functional group

chloro
ester

storage temp.

2-8°C

SMILES string

COC(=O)C(=C)CCl

InChI

1S/C5H7ClO2/c1-4(3-6)5(7)8-2/h1,3H2,2H3

InChI key

NYMDTEIPYQNXIL-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.0 °F

Flash Point(C)

62.77 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service