Skip to Content
Merck
All Photos(1)

Key Documents

713910

Sigma-Aldrich

2-(4,5-Dihydro-2-oxazolyl)quinoline

97%

Synonym(s):

2-(2-Quinolinyl)oxazoline, Sigman ligand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10N2O
CAS Number:
Molecular Weight:
198.22
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥96.5% (HPLC)
97%

form

solid

reaction suitability

reagent type: ligand
reaction type: Cross Couplings

SMILES string

C1CN=C(O1)c2ccc3ccccc3n2

InChI

1S/C12H10N2O/c1-2-4-10-9(3-1)5-6-11(14-10)12-13-7-8-15-12/h1-6H,7-8H2

InChI key

USDSJWOYSHFPND-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service