Skip to Content
Merck
All Photos(1)

Key Documents

712582

Sigma-Aldrich

O-[N-(3-Maleimidopropionyl)aminoethyl]-O′-[3-(N-succinimidyloxy)-3-oxopropyl]triethylene glycol

≥90% (NMR)

Synonym(s):

N-[15-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-15-oxo-3,6,9,12-tetraoxapentadec-1-yl]-2,5-dihydro-2,5-dioxo-1H-pyrrole-1-propanamide, monodisperse Mal-PEG4-NHS-ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31N3O11
CAS Number:
Molecular Weight:
513.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥90% (NMR)

form

liquid

reaction suitability

reagent type: cross-linking reagent

functional group

NHS ester
maleimide

storage temp.

−20°C

SMILES string

O=C(NCCOCCOCCOCCOCCC(ON1C(CCC1=O)=O)=O)CCN2C(C=CC2=O)=O

InChI

1S/C22H31N3O11/c26-17(5-8-24-18(27)1-2-19(24)28)23-7-10-33-12-14-35-16-15-34-13-11-32-9-6-22(31)36-25-20(29)3-4-21(25)30/h1-2H,3-16H2,(H,23,26)

InChI key

XSRYGQJQNPJNKZ-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Circulatory half-life is a key success factor for new drugs. In this respect, PEGylation or PEG-ing—the modification of potential candidates ranging from non-peptidic small molecules to peptides and proteins, antibody fragments, aptamers, and saccharides or oligonucleotides with polyethylene glycol chains—offers numerous advantages.

Circulatory half-life is a key success factor for new drugs. In this respect, PEGylation or PEG-ing—the modification of potential candidates ranging from non-peptidic small molecules to peptides and proteins, antibody fragments, aptamers, and saccharides or oligonucleotides with polyethylene glycol chains—offers numerous advantages.

Circulatory half-life is a key success factor for new drugs. In this respect, PEGylation or PEG-ing—the modification of potential candidates ranging from non-peptidic small molecules to peptides and proteins, antibody fragments, aptamers, and saccharides or oligonucleotides with polyethylene glycol chains—offers numerous advantages.

Circulatory half-life is a key success factor for new drugs. In this respect, PEGylation or PEG-ing—the modification of potential candidates ranging from non-peptidic small molecules to peptides and proteins, antibody fragments, aptamers, and saccharides or oligonucleotides with polyethylene glycol chains—offers numerous advantages.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service