Skip to Content
Merck
All Photos(1)

Key Documents

696447

Sigma-Aldrich

N-Boc-piperidine-4-carbonitrile

97%

Synonym(s):

1,1-Dimethylethyl 4-cyano-1-piperidinecarboxylate, 1-Boc-4-cyanopiperidine, 4-Cyano-1-piperidinecarboxylic acid 1,1-dimethylethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18N2O2
CAS Number:
Molecular Weight:
210.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

powder

mp

60-63 °C

functional group

nitrile

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CCC(CC1)C#N

InChI

1S/C11H18N2O2/c1-11(2,3)15-10(14)13-6-4-9(8-12)5-7-13/h9H,4-7H2,1-3H3

InChI key

UQADQTBQNVARAP-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Aminomethylated fluoropiperidines
  • Protein kinase B inhibitors
  • GlyT1 inhibitors
  • Piperidinecarboxylic acids via nitrilase-catalyzed enantioselective synthesis

Reactant for
  • Replacement in CB2 receptor inhibitors
  • Double addition reactions of methyllithium and n-butyllithium to unsaturated nitriles

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service