Skip to Content
Merck
All Photos(3)

Key Documents

686670

Sigma-Aldrich

Polyester-8-hydroxyl-1-carboxyl bis-MPA dendron, generation 3

≥97%

Synonym(s):

Dendron-G3-Carboxyl-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H58O22
Molecular Weight:
830.82
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

hydroxyl surface groups

Quality Level

Assay

≥97%

form

powder (lyophilized)

mol wt

generation 3

feature

focal point carboxylic acid

no. Surface Groups

8

storage temp.

2-8°C

SMILES string

CC(CO)(CO)C(=O)OCC(C)(COC(=O)C(C)(CO)CO)C(=O)OCC(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)C(O)=O

InChI

1S/C35H58O22/c1-29(8-36,9-37)23(46)54-18-34(6,19-55-24(47)30(2,10-38)11-39)27(50)52-16-33(5,22(44)45)17-53-28(51)35(7,20-56-25(48)31(3,12-40)13-41)21-57-26(49)32(4,14-42)15-43/h36-43H,8-21H2,1-7H3,(H,44,45)

InChI key

CSKGZAJBJPVHFW-UHFFFAOYSA-N

General description

Soluble in a variety of polar solvents, such as DMF, methanol, and water.
calculated mol. wt. 831

Application

Manufactured by Polymer Factory Sweden AB.

Other Notes

Bis-MPA = 2,2-Bis(hydroxyl-methyl)propionic acid

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Dendrons in nanomaterials - Find features and benefits of polyester dendrons and hyperbranched polymer products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service